
Sodium periodate
Catalog # 104085-812
Supplier: Hach
CAS Number:
Specifications
- Cat. No.:104085-812
Specifications
About this item
Merck Index:13,08714
Boiling Pt:300 °C (1013 hPa)
Melting Pt:175 °C
Density:3.87 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:5.1,II
:213.89 g/mol
EINECS:232-197-6
UN:1479
:GHS03
CAS Number:7790-28-5
:
Formula:NaIO₄
MDL Number:MFCD00003534