Order Entry
United States
ContactUsLinkComponent
Ferroin (1,10-phenanthrolineferrous sulfate complex) solution indicator, for COD determination
Ferroin (1,10-phenanthrolineferrous sulfate complex) solution indicator, for COD determination
Catalog # 104085-616
Supplier:  Hach
CAS Number:  
undefined
Ferroin (1,10-phenanthrolineferrous sulfate complex) solution indicator, for COD determination
Catalog # 104085-616
Supplier:  Hach
Supplier Number:  181233
CAS Number:  

Specifications

  • Cat. No.:
    104085-616

Specifications

About this item

 :692.54
CAS Number:14634-91-4
 :
Formula:C₃₆H₂₄FeN₆SO₄
MDL Number:MFCD00036428