Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
tri-Sodium citrate dihydrate
104085-578
:
Hach
:
tri-Sodium citrate dihydrate
104085-578
:
Hach
:
1490849
:
Cat. No.:
104085-578
:
294.1 g/mol
:
13,08675
:
150 °C
:
1.76 g/cm³ (20 °C)
:
Ambient
:
200-675-3
:
6132-04-3
:
:
Na₃C₆H₅O₇·2H₂O
:
MFCD00150031