Order Entry
United States
ContactUsLinkComponent
 
Magnesium chloride 510 g/L in aqueous solution
Catalog # 104085-550
Supplier:  Hach
CAS Number:  
Magnesium chloride 510 g/L in aqueous solution
Catalog # 104085-550
Supplier:  Hach
Supplier Number:  1476253
CAS Number:  

Specifications

  • Cat. No.:
    104085-550

Specifications

About this item

 :95.21 g/mol
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7786-30-3
 :
Formula:MgCl₂
MDL Number:MFCD00011106