Orders LinkContactUsLinkComponent
Sample Cuvettes
Marketsource
Supplier:   HACH COMPANY MS
Sample Cuvettes
Supplier:  HACH COMPANY MS
Marketsource
Certificates
Sample Cuvettes
8 Options Available Below
Product Details & Documents
Sample cuvettes for photometric measurements.
  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Product Family Options
Square
Product Information
  • Pathlength
    Window material
    Availability
    Price
  • 10 mm
    Quartz glass
    Log in for availability
    Log in for availability
    Each
    Each - $534.46
    $534.46
    Each
       $534.46
    Supplier Number: 2624410
    Specifications:
    Marketsource
    Marketsource
  • 10 mm
    Quartz glass
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $916.62
    $916.62
    Pack of 2
       $916.62
    Supplier Number: 4822800
    Specifications:
    Marketsource
    Marketsource
  • 10 mm
    Optical glass
    Log in for availability
    Log in for availability
    Pack of 3
    Pack of 3 - $220.15
    $220.15
    Pack of 3
       $220.15
    Supplier Number: LZP045
    Specifications:
    Marketsource
    Marketsource
  • 1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $411.23
    $411.23
    Pack of 2
       $411.23
    Supplier Number: 2495402
    Specifications:
    Marketsource
    Marketsource
  • 1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    Pack of 8
    Pack of 8 - $1,168.62
    $1,168.62
    Pack of 8
       $1,168.62
    Supplier Number: 2495408
    Specifications:
    Marketsource
    Marketsource
Rectangular
Product Information
  • Pathlength
    Window material
    Availability
    Price
  • 50 mm
    Glass
    Log in for availability
    Log in for availability
    Each
    Each - $412.62
    $412.62
    Each
       $412.62
    Supplier Number: 2629250
    Specifications:
    Marketsource
    Marketsource
Round
Product Information
  • Pathlength
    Window material
    Availability
    Price
  • 13 mm
    Glass
    Discontinued
    Supplier Number: LCW906
    Specifications:
    Marketsource
    Marketsource
  • 1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    Each
    Each - $136.51
    $136.51
    Each
       $136.51
    Supplier Number: 2401906
    Specifications:
    Marketsource
    Marketsource
  • 1" / 25 mm
    Glass
    Log in for availability
    Log in for availability
    Each
    Each - $21.03
    $21.03
    Each
       $21.03
    Supplier Number: 2122800
    Specifications:
    Marketsource
    Marketsource