Order Entry
United States
ContactUsLinkComponent
Baseline® Aneroid Sphygmomanometer Unit
Baseline® Aneroid Sphygmomanometer Unit
Catalog # 470357-422
New Product
Baseline® Aneroid Sphygmomanometer Unit
Catalog # 470357-422
Supplier Number:  12-2250
New Product

Specifications

  • Color:
    Black
  • Description:
    Aneroid sphygmomanometer unit
  • Cat. No.:
    470357-422

Specifications

About this item

Baseline aneroid sphygmomanometer unit for blood pressure unit.

  • Circumference: 23 to 40 cm
  • Cuff material: Nylon cuff
  • Display type: Dial display
  • Number of tubes: 2-tubes
  • Size: Adult cuff
  • Style: Pocket aneroid
  • Target area: Arm
  • Usage: Reusable