Order Entry
United States
ContactUsLinkComponent
Phosphate Buffered Saline (PBS) 1X with 0.05% Tween 20, pH 7.4
Phosphate Buffered Saline (PBS) 1X with 0.05% Tween 20, pH 7.4
Catalog # 77442-720
Supplier:  Teknova
CAS Number:  
Phosphate Buffered Saline (PBS) 1X with 0.05% Tween 20, pH 7.4
Catalog # 77442-720
Supplier:  Teknova
Supplier Number:  P0276
CAS Number:  

Specifications

  • Size:
    20 L
  • Shelf Life:
    1095 days
  • Storage Temperature:
    Store at room temperature
  • Cat. No.:
    77442-720
  • pH:
    7.40 ±0.04

Specifications

About this item

1X Solution composition: 137 mM Sodium chloride, 2.7 mM potassium chloride, 4.3 mM sodium phosphate, 1.4 mM potassium phosphate and 0.05% Tween (v/v).