Order Entry
United States
ContactUsLinkComponent
Phosphate Buffered Saline (PBS) 1X with 0.1% Tween 80
Phosphate Buffered Saline (PBS) 1X with 0.1% Tween 80
Catalog # 77442-728
Supplier:  Teknova
CAS Number:  
Phosphate Buffered Saline (PBS) 1X with 0.1% Tween 80
Catalog # 77442-728
Supplier:  Teknova
Supplier Number:  P0215
CAS Number:  

Specifications

  • Size:
    1000 ml
  • Shelf Life:
    1095 days
  • Storage Temperature:
    Store at room temperature
  • Cat. No.:
    77442-728

Specifications

About this item

PBS 1X Solution composition: 137.0 mM Sodium chloride, 4.3 mM sodium phosphate dibasic, 2.7 mM potassium chloride, 1.4 mM potassium phosphate monobasic and 0.1% Tween 80.