ContactUsLinkComponent
AlerTox® Milk Lateral Flow Test Sticks
AlerTox® Milk Lateral Flow Test Sticks
Catalog # 77661-324
Supplier:  Hardy Diagnostics
New Product
CAS Number:  
AlerTox® Milk Lateral Flow Test Sticks
Catalog # 77661-324
Supplier:  Hardy Diagnostics
Supplier Number:  KIT3015
New Product
CAS Number:  
Specifications
  • Description:
    Milk lateral flow test sticks
  • Cat. No.:
    77661-324
  • No. of strips:
    10
Specifications
About this item
Designed specifically to help manufacturers keep items free of antigens that may cause allergic reactions.
  • Quick and reliable
  • No cross-reactivity
  • Results in 10 minutes
  • High sensitivity
AlerTox® Sticks are a rapid, user-friendly test for the detection and qualification of main food allergens with no lab equipment or scientific knowledge required.