

Sodium nitrate ≥99.99% (trace metals basis), SPEX Infinitas™
Supplier: SPEX CERTIPREP LLC
New Product
Certificates
About this item
CAS Number:7631-99-4
:
Formula:NaNO₃
MDL Number:MFCD00011119
:84.99 g/mol
Boiling Pt:600...700 °C (1112...1292 °F)
Melting Pt:312 °C (593.6 °F)
Density:2.26 g/cm³ (18.8597 lbs./gal)
UN:1498
ADR:5.1,III
:GHS03
:GHS07
Frequently Bought Together