HICAL Acids Mix
Catalog # SXHICAL-ACIDS
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Cat. No.:SXHICAL-ACIDS
- Storage temperature:Freezer
Specifications
About this item
Semivolatiles, US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.
CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.