Fortification Solution
SX5242-F
New Product
:
- Cat. No.:SX5242-F
- Storage temperature:Ambient
US EPA method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to methanol 524.3 revision 4 are supplied in a 24-component mix.