Purgeable Gases Mix B
Catalog # SX624-BH
Supplier: SPEX CERTIPREP LLC
New Product
CAS Number:
Specifications
- Cat. No.:SX624-BH
- Storage temperature:Freezer
Specifications
About this item
US EPA method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.