Undercounter Flammables Cabinets, SECURALL®
Supplier: Securall Cabinets
Certificates
About this item
Avoid the fire and explosion hazards of unprotected flammable liquids and chemicals with laboratory storage cabinets.
- 18-gauge galvanized steel construction
- One galvanized shelf and flush mounted handle
Cabinets are designed to fit under most counters, giving more workspace on countertops. The cabinets feature has kick recess, and a grounding connection located in the back of the cabinet. The label reads 'FLAMMABLE – KEEP FIRE AWAY'.
Color: Light neutral
Accessories information: .
Ordering information: This item is considered a Standard Custom Item built to VWR® Specification, and, as such, cannot be returned.
Specifications
- Catalog No:
- CFBRC-FLAM-36
- CFBRC-FLAM-24
- Environmentally Preferable:
- Sustainable Materials
- Sustainable Materials
- Environmental Certification:
- SMaRT Certified
- SMaRT Certified