To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Species:Human
- Description:CellAdhere™ Dilution buffer
- Size:100 ml
- Cat. No.:MSPP-07183
Specifications
About this item
Dilution buffer for matrix proteins.
Use CellAdhere™ Dilution buffer in combination with Vitronectin XF™ cell culture matrix (developed and manufactured by Nucleus Biologics) to provide a defined, xeno-free surface for the culture of human embryonic stem (ES) and induced pluripotent stem (iPS) cells.
CellAdhere™ is required for use with Vitronectin XF™, which may be used as an alternative to Corning® Matrigel® to support the growth and differentiation of human pluripotent stem cells.