Order Entry
United States
ContactUsLinkComponent
BD BBL™ Saline 0.9% Tube, BD Diagnostics
BD BBL™ Saline 0.9% Tube, BD Diagnostics
Catalog # 90008-928
Supplier:  BD
CAS Number:  
BD BBL™ Saline 0.9% Tube, BD Diagnostics
Catalog # 90008-928
Supplier:  BD
Supplier Number:  215128
CAS Number:  

Specifications

  • Packed:
    100 tubes/case
  • Cat. No.:
    90008-928
  • Format:
    Tube
  • Volume:
    9 ml
  • Supplier no.:
    215128

Specifications

About this item

Saline 0.9%, in 9 ml tubes. 100 tubes per case.