ContactUsLinkComponent
Logic Remote Communication Board
Logic Remote Communication Board
  77574-622
 :  Labconco
New Product
undefined
Logic Remote Communication Board
  77574-622
 :  Labconco
 :  3289000
New Product
  • Description:
    Logic remote communication board
  • Cat. No.:
    77574-622
Logic remote communication board for connecting biosafety cabinet to remote exhaust blower or building monitoring system.
  • For Type A2 cabinets
  • Shipping weight 0.5 kg (1.0 lbs.)
This board is factory installed on Logic Type B2 Cabinets. A2 Cabinets require installation.
 : Includes three dry contact relays: USB data output, RS232 RJ45 data output and an ethernet pass-through.