Orders LinkContactUsLinkComponent
EasySep™ Release Human CD19 Positive Selection Kit
EasySep™ Release Human CD19 Positive Selection Kit
EasySep™ release human CD19 positive selection kit, 1×10⁹ cells
Catalog # MSPP-17754
Marketsource
Supplier #:  17754
EasySep™ Release Human CD19 Positive Selection Kit
Catalog # MSPP-17754
Marketsource
EasySep™ Release Human CD19 Positive Selection Kit
EasySep™ Release Human CD19 Positive Selection Kit
EasySep™ release human CD19 positive selection kit, 1×10⁹ cells
Price
$1,361.54
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Immunomagnetic positive selection of particle-free human CD19+ cells.
  • Highly purified human CD19+ cells isolated in less than 30 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Human CD19 Positive Selection Kit is designed to isolate CD19+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection.
Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD19+ cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands. The CD19 antigen is expressed on mature B cells and B cell progenitors but is lost upon maturation to plasma cells. It is also expressed on follicular dendritic cells.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ release human CD19 positive selection kit