ContactUsLinkComponent
EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit
EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit
Catalog # MSPP-17861
undefined
EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit
Catalog # MSPP-17861
Supplier Number:  17861
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    1×10⁹ cells
  • Description:
    EasySep™ human Pan-CD25 positive selection and depletion Kit
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-17861
Specifications
About this item
Immunomagnetic depletion of human CD25+ cells.
  • Fast, easy-to-use and column-free
  • Up to 1.3 log depletion of CD25+ cells
  • Up to 98% purity
The EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit is designed to deplete CD25+ cells from fresh peripheral blood mononuclear cells or lysed leukapheresis samples. CD25+ cells are targeted with Tetrameric Antibody Complexes recognizing CD25 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off. The CD25 antigen is expressed on regulatory T cells and activated T and B cells.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').