Specifications
- Capacity:2×10⁹ cells
- Description:EasySep™ mouse CD11b positive selection kit II
- Environmentally Preferable:
- Cat. No.:MSPP-18970
Specifications
About this item
Immunomagnetic positive selection of CD11b+ cells (e.g. monocytes) from mouse splenocytes or other tissues.
- Fast, easy-to-use, and column-free
- Up to 95% purity (for bone marrow, purity can be up to 99%)
- Isolated cells are not fluorochrome-labeled
The EasySep™ Mouse CD11b Positive Selection Kit II isolates highly purified CD11b+ cells from mouse splenocytes, bone marrow, lungs, brains or other tissues by immunomagnetic positive selection. Desired cells are targeted with antibodies and magnetic particles, and isolated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated cells are immediately ready for downstream applications such as flow cytometry, culture, and cell-based experiments.
This product replaces the EasySep™ Mouse CD11b Positive Selection Kit for even faster cell isolations and does not result in the labeling of isolated cells with PE.