Orders LinkContactUsLinkComponent
EasySep™ Direct Human CD8+ T Cell Isolation Kit
EasySep™ Direct Human CD8+ T Cell Isolation Kit
EasySep™ direct human CD8+ T cell isolation kit, 100 ml whole blood processed
Catalog # MSPP-19663
Marketsource
Supplier #:  19663
EasySep™ Direct Human CD8+ T Cell Isolation Kit
Catalog # MSPP-19663
Marketsource
EasySep™ Direct Human CD8+ T Cell Isolation Kit
EasySep™ Direct Human CD8+ T Cell Isolation Kit
EasySep™ direct human CD8+ T cell isolation kit, 100 ml whole blood processed
Price
$927.69
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Immunomagnetic isolation of CD8+ T cells directly from whole blood
  • >99.9% red blood cell (RBC) depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 91% purity of isolated cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human CD8+ T Cell Isolation Kit is designed to isolate CD8+ T cells from human whole blood by immunomagnetic negative selection. The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood with antibody complexes and magnetic particles. The magnetically labeled RBCs, platelets and cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
  • Capacity:
    100 ml whole blood processed
  • Description:
    EasySep™ direct human CD8+ T cell isolation kit