EasySep™ Direct Human NK Cell Isolation Kit
MSPP-19665
- Capacity:100 ml whole blood processed
- Description:EasySep™ direct human NK cell isolation kit
- Environmentally Preferable:
- Cat. No.:MSPP-19665
Immunomagnetic isolation of NK cells directly from whole blood
- >99.9% red blood cell (RBC) depletion without the need for density gradient centrifugation, sedimentation, or lysis
- Up to 84% purity of isolated cells
- Fast, easy-to-use and column-free
- Isolated cells are untouched
The EasySep™ Direct Human NK Cell Isolation Kit is designed to isolate NK cells from human whole blood by immunomagnetic negative selection. The EasySep™ Direct procedure involves labeling RBCs, platelets and unwanted cells present in human whole blood with antibody complexes and magnetic particles. The magnetically labeled RBCs, platelets and cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').