ContactUsLinkComponent
Secchi Disc
Secchi Disc
Catalog # 104085-888
Supplier:  Hach
undefined
Secchi Disc
Catalog # 104085-888
Supplier:  Hach
Supplier Number:  2177200
Specifications
  • Description:
    Secchi disc
  • Cat. No.:
    104085-888
Specifications
About this item
The disc is designed to measure water transparency in situ via the semi-quantitative method.
  • Disc slowly lowered in the water until the pattern is no longer visible
  • Depth of immersion is then evaluated
  • Measure related to the transparency
For use in oceans and lakes.
Ø = 200 mm
Certifications: ISO 7027.