ContactUsLinkComponent
EasySep™ Mouse F4/80 Positive Selection Kit
EasySep™ Mouse F4/80 Positive Selection Kit
Catalog # MSPP-100-0659
undefined
EasySep™ Mouse F4/80 Positive Selection Kit
Catalog # MSPP-100-0659
Supplier Number:  100-0659
Specifications
  • Capacity:
    2×10⁹ cells
  • Description:
    EasySep™ Mouse F4/80 positive selection kit
  • Cat. No.:
    MSPP-100-0659
Specifications
About this item
Immunomagnetic positive selection of F4/80+ cells from mouse splenocytes, lung tissue, and peritoneal lavage.
  • Fast, easy-to-use, and column-free
  • Up to 90% purity
  • Isolated cells are not fluorochrome-labeled
Easily isolate F4/80+ macrophages in as little as 24 minutes—without the use of columns—from mouse splenocytes, lung tissue, and peritoneal lavage.
Using immunomagnetic positive selection, F4/80+ cells are labeled with antibodies and dextran-coated magnetic particles (RapidSpheres™), for column-free cell separation using an EasySep™ magnet. Isolated cells are highly purified and ready for your downstream applications.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).