EasySep™ Human CD4 Positive Selection Kit II
About this item
Immunomagnetic positive selection of human CD4+ cells (e.g. CD4+ T cells).
- Fast and easy-to-use
- Up to 96% purity
- No columns required
The EasySep™ Human CD4 Positive Selection Kit II is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognizing CD4 and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human CD4 Positive Selection Kit (STMC18052) for even faster cell isolations.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Immunomagnetic positive selection of human CD4+ cells (e.g. CD4+ T cells).
- Fast and easy-to-use
- Up to 96% purity
- No columns required
The EasySep™ Human CD4 Positive Selection Kit II is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection. Desired cells are targeted with antibody complexes recognizing CD4 and magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This product replaces the EasySep™ Human CD4 Positive Selection Kit (STMC18052) for even faster cell isolations.
: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Product Family Options
Product Information
- CapacityAvailabilityPrice
- MSPP-17852EasySep™ human CD4 positive selection kit IISupplier Number: 17852Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- MSPP-100-0693EasySep™ human CD4 positive selection kit IISupplier Number: 100-0693Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- MSPP-17852RFEasySep™ kit with RoboSep™ buffer and RoboSep™ filter tipsEasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips1×10⁹ cellsLog in for availabilityEach$1,373.85Supplier Number: 17852RFItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource