Linearity FLQ Vitamin D
Catalog # 77627-800
Supplier: Audit Microcontrols
New Product
CAS Number:
Specifications
- Pack type:5 vials
- Description:Linearity FLQ Vitamin D
- Size:5×2 ml
- Storage Conditions:–15 °C
- Storage Temperature:2...8 °C once thawed
- Cat. No.:77627-800
- Linearity:Linearity
- Stability:Once thawed, store at 2...8 °C for 14 days
- Analyte:Vitamin D
- Volume:2 ml
Specifications
About this item
Linearity FLQ Vitamin D is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analyte: Vitamin D.
Certifications: CE marked.