Order Entry
United States
ContactUsLinkComponent
Rat VIP ELISA Kit
Rat VIP ELISA Kit
Catalog # MSPP-CEA380RA
CAS Number:  
undefined
Rat VIP ELISA Kit
Catalog # MSPP-CEA380RA
Supplier Number:  CEA380RA
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Rat
  • Target protein:
    VIP
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Vasoactive Intestinal Peptide (VIP) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Shelf Life:
    12 Months
  • Detection Range:
    6.17 - 500 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    50 µl
  • Sensitivity:
    2.58 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-CEA380RA
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Rat VIP (Vasoactive Intestinal Peptide). The assay range is from 6.17 to 500 pg/ml (Competitive kit) with a sensitivity of 2.58 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Vasoactive intestinal peptide (VIP) is a peptide hormone containing 28 amino acid residues and is produced in many areas of the human body including the gut, pancreas and suprachiasmatic nuclei of the hypothalamus in the brain.It has a half-life in the blood of about two minutes. With respect to the digestive system, VIP seems to induce smooth muscle relaxation (lower esophageal sphincter, stomach, gallbladder), stimulate secretion of water into pancreatic juice and bile, and cause inhibition of gastric acid secretion and absorption from the intestinal lumen. Its role in the intestine is to greatly stimulate secretion of water and electrolytes, as well as dilating intestinal smooth muscle, dilating peripheral blood vessels, stimulating pancreatic bicarbonate secretion, and inhibiting gastrin-stimulated gastric acid secretion.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.