- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Mouse
- Target protein:REN
- Size:1 Kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Renin (REN) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:15.6 - 1000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:6.1 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA889MU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Mouse REN (Renin). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Renin is a member of the aspartyl proteinase family produced largely in part by the juxtaglomerular cells in the kidney. Renin differs from the other members of this class by having a pH optimum near neutral pH region with native substrates instead of 2.0 to 3.4 This more neutral pH optimum allows it to be functional in the plasma. Renin also has very high selectivity for substrate due to quite long peptide recognition on either side of the peptide bond undergoing cleavage. An octapeptide substrate was the minimum length to be cleaved by renin. Renin plays a crucial role in the regulation of blood pressure and salt balance through the cleavage of angiotensinogen, which is the only known physiological substrate of renin.