Order Entry
United States
ContactUsLinkComponent
Mouse NT3 ELISA Kit
Mouse NT3 ELISA Kit
Catalog # MSPP-SEA106MU
CAS Number:  
undefined
Mouse NT3 ELISA Kit
Catalog # MSPP-SEA106MU
Supplier Number:  SEA106MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    NT3
  • Size:
    1 Kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Neurotrophin 3 (NT3) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    78 - 5000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    28 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA106MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse NT3 (Neurotrophin 3). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 28 pg/ml. There is no detectable cross to reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Neurotrophin-3,or NT-3 is a protein that  is encoded by the NTF3 gene, NT-3 is a neurotrophic factor, in the NGF (Nerve Growth Factor)-family of neurotrophins. It is a protein growth factor which has activity on certain neurons of the peripheral and central nervous system; it helps to support the survival and differentiation of existing neurons, and encourages the growth and differentiation of new neurons and synapses. NT-3 was the third neurotrophic factor to be characterized, after nerve growth factor (NGF) and BDNF (Brain Derived Neurotrophic). NT-3 is unique in the number of neurons it can potentially stimulate, given its ability to activate two of the receptor tyrosine kinase neurotrophin receptors (TrkC and TrkB).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.