Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse D2D ELISA Kit

Mouse D2D ELISA Kit

96 wells, 1 Kit
Catalog # MSPP-CEA506MU
Marketsource
Supplier:   Cloud-Clone
Supplier #:  CEA506MU
Mouse D2D ELISA Kit
Catalog # MSPP-CEA506MU
Supplier:  Cloud-Clone
Marketsource
CAS Number:  
Mouse D2D ELISA Kit
Mouse D2D ELISA Kit

96 wells, 1 Kit

CAS Number:  
Price
$576.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This assay has high sensitivity and excellent specificity for detecting Mouse D2D (D to Dimer). The assay range is from 617.3 to 50,000 pg/ml (Competitive kit) with a sensitivity of 243.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

D-dimers are unique in that they are the breakdown products of a fibrin mesh that has been stabilized by Factor XIII. This factor crosslinks the E-element to two D-elements. This is the final step in the generation of a thrombus. Plasmin is a fibrinolytic enzyme that organizes clots and breaks down the fibrin mesh. It cannot, however, break down the bonds between one E and two D units. The protein fragment thus left over is a D-dimer. D-dimer concentration may be determined by a blood test to help diagnose thrombosis. Since its introduction in the 1990s, it has become an important test performed in patients suspected of thrombotic disorders. While a negative result practically rules out thrombosis, a positive result can indicate thrombosis but does not rule out other potential causes. Its main use, therefore, is to exclude thromboembolic disease where the probability is low.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.

Documents

Specifications

Specifications

  • Size:
    1 Kit
  • No. of tests:
    96 wells
  • Primary antibody reactivity:
    Mouse
  • Detection Range:
    617.3 - 50000 pg/ml
  • Sample Volume:
    50 µl
  • Host:
    Rabbit
  • Assay Type:
    Competitive
  • Time to Results:
    2 h
  • Detection Method:
    Colorimetric
  • Cat. No.:
    MSPP-CEA506MU
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    243.7 pg/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Target protein:
    D2D
  • Sample Type:
    Plasma
  • Shelf Life:
    12 Months
  • Cross Reactivity:
    No significant cross-reactivity or interference between D-Dimer (D2D) and analogues was observed
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year

Specifications