Order Entry
United States
ContactUsLinkComponent
Human IL1b ELISA Kit
Human IL1b ELISA Kit
Catalog # MSPP-SEA563HU
CAS Number:  
undefined
Human IL1b ELISA Kit
Catalog # MSPP-SEA563HU
Supplier Number:  SEA563HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL1b
  • Size:
    1 Kit
  • Sample Type:
    Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 1 Beta (IL1b) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.1 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA563HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human IL1b (Interleukin 1 Beta). The assay range is from 15.6 to 1,000 pg/ml (Sandwich kit) with a sensitivity of 6.1 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-Assay: CV <10%, Inter-Assay: CV <12%

Interleukin-1 (IL-1) is one of the first cytokines ever described. Its initial discovery was as a factor that could induce fever, control lymphocytes, increase the number of bone marrow cells and cause degeneration of bone joints. At this time, IL-1 was known under several other names including endogenous pyrogen, lymphocyte activating factor, haemopoetin-1 and mononuclear cell factor, amongst others. It was around 1984 to 1985 when scientists confirmed that IL-1 was actually composed of two distinct proteins, now called IL-1α and IL-1β. These belong to a family of cytokines known as the interleukin-1 superfamily.Both IL-1α and IL-1β are produced by macrophages, monocytes and dendritic cells.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.