- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:IL10
- Size:1 Kit
- Sample Type:Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between High Sensitive Interleukin 10 (IL10) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:1.56 - 100 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.64 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-HEA056HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human IL10 (Interleukin 10). The assay range is from 1.56 to 100 pg/ml (Sandwich kit) with a sensitivity of 0.64 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-Assay: CV <10%, Inter-Assay: CV <12%
Interleukin-10 is a small protein known as a cytokine that functions as an important regulator of the immune system. Although IL-10 is known to have many different roles in the immune system, its two major activities include inhibition of cytokine production by macrophages and inhibition of the accessory functions of macrophages during T cell activation. The effects of these actions cause IL-10 to play mainly an anti-inflammatory role in the immune system. IL-10 was originally known as the cytokine synthesis inhibiting factor (CSIF), and the discovery of this protein was based on its biological activity. IL-10 is mainly produced by the Th2 subset of CD4+ helper cells. However, it is also produced by some activated B cells, some Th1 cells, activated macrophages, and some nonhematopoietic sources.