Order Entry
United States
ContactUsLinkComponent
Mouse IL5 ELISA Kit
Mouse IL5 ELISA Kit
Catalog # MSPP-SEA078MU
CAS Number:  
undefined
Mouse IL5 ELISA Kit
Catalog # MSPP-SEA078MU
Supplier Number:  SEA078MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL5
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Interleukin 5 (IL5) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    6.0 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA078MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse IL5 (Interleukin 5). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 6.0 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

IL-5 is secreted by a restricted number of mesenchymal cell types. In its native state, mature IL-5 is synthesized as a 115 aa, highly glycosylated 22 kDa monomer that forms a 40 to 50 kDa disulfide-linked homodimer. Although the content of  carbohydrate  is  high, carbohydrate is not needed for bioactivity.  Monomeric IL-5 has no activity and requires a homodimer for function, which is in contrast to the receptor-related IL-3 and GM-CSF cytokines that exist only as monomers.  Just as one IL-3 and GM-CSF monomer binds to one receptor, one IL-5 homodimer is able to engage only one IL-5 receptor.  It has been suggested that IL-5 (as a dimmer) undergoes a general conformational change after binding to one receptor molecule and this change precludes binding to a second receptor.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.