Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Mouse
- Target protein:GDNF
- Size:1 kit
- Sample Type:Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Glial Cell Line Derived Neurotrophic Factor (GDNF) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:78 - 5000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:32 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEA043MU
- No. of tests:96 wells
Specifications
About this item
This assay has high sensitivity and excellent specificity for detecting Mouse GDNF (Glial Cell Line Derived Neurotrophic Factor). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 32 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Glial Cell Line-derived Neurotrophic Factor (GDNF) is a highly conserved neurotrophic factor. The recombinant form of this protein was shown to promote the survival and differentiation of dopaminergic neurons in culture, and was able to prevent apoptosis of motor neurons induced by axotomy. The protein is processed to a mature secreted form that exists as a homodimer. The mature form of the protein is a ligand for the product of the RET (rearranged during transfection) protooncogene. In addition to the transcript encoding GDNF, two additional alternative transcripts encoding distinct proteins, referred to as astrocyte-derived trophic factors, have also been described. Mutations in this gene may be associated with Hirschsprung disease.