Order Entry
United States
ContactUsLinkComponent
Mouse CFH ELISA Kit
Mouse CFH ELISA Kit
Catalog # MSPP-SEA635MU
CAS Number:  
undefined
Mouse CFH ELISA Kit
Catalog # MSPP-SEA635MU
Supplier Number:  SEA635MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    CFH
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Complement Factor H (CFH) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    7.8 - 500 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    3.7 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA635MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse CFH (Complement Factor H). The assay range is from 7.8 to 500 ng/ml (Sandwich kit) with a sensitivity of 3.7 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Factor H is a member of the regulators of complement activation family and is a complement control protein. It is a large (155 kilodaltons), soluble glycoprotein that circulates in human plasma (at a concentration of 500–800 micrograms per milliliter). Its main job is to regulate the Alternative Pathway of the complement system, ensuring that the complement system is directed towards pathogens and does not damage host tissue. Factor H regulates complement activation on self cells by possessing both cofactor activity for the Factor I mediated C3b cleavage, and decay accelerating activity against the alternative pathway C3 convertase, C3bBb. Factor H protects self cells from complement activation but not bacteria/viruses, in that it binds to glycosaminoglycans (GAGs) that are present on host cells but not pathogen cell surfaces.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.