Order Entry
United States
ContactUsLinkComponent
Mouse ACE2 ELISA Kit
Mouse ACE2 ELISA Kit
Catalog # MSPP-SEB886MU
CAS Number:  
undefined
Mouse ACE2 ELISA Kit
Catalog # MSPP-SEB886MU
Supplier Number:  SEB886MU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    ACE2
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Angiotensin I Converting Enzyme 2 (ACE2) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    62.5 - 4000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    26.5 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB886MU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Mouse ACE2 (Angiotensin I Converting Enzyme 2). The assay range is from 62.5 to 4000 pg/ml (Sandwich kit) with a sensitivity of 26.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

ACE2 is an exopeptidase that catalyses the conversion of angiotensin I to the nonapeptide angiotensin,or the conversion of angiotensin II to angiotensin. ACE 2 has direct effects on cardiac function, and is expressed predominantly in vascular endothelial cells of the heart and the kidneys.By EST database searching for sequences showing homology to the zinc metalloprotease angiotensin-I converting enzyme (ACE) and by screening a human lymphoma cDNA library, Tipnis et al., (2000) cloned a full-length ACE2 cDNA, which they called ACEH, encoding a deduced 805-amino acid protein that shares approximately 40% identity with the N- and C-terminal domains of ACE. ACE2 contains a potential 17-amino acid N-terminal signal peptide and a putative 22-amino acid C-terminal membrane anchor.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.