Order Entry
United States
ContactUsLinkComponent
Human WNT10B ELISA Kit
Human WNT10B ELISA Kit
Catalog # MSPP-SEP553HU
CAS Number:  
undefined
Human WNT10B ELISA Kit
Catalog # MSPP-SEP553HU
Supplier Number:  SEP553HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    WNT10B
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Wingless Type MMTV Integration Site Family, Member 10B (WNT10B) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.312 - 20 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.114 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEP553HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human WNT10B (Wingless Type MMTV Integration Site Family, Member 10B). The assay range is from 0.312 to 20 ng/ml (Sandwich kit) with a sensitivity of 0.114 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

WNT10B is a member of the WNT gene family. It may be involved in breast cancer, and its protein signaling is likely a molecular switch that governs adipogenesis. This protein is 96% identical to the mouse Wnt10b protein at the amino acid level. This gene is clustered with another family member, WNT1, in the chromosome 12q13 region. The human WNT10B sequence is 88% and 95% identical to the mouse gene at nucleotide and amino acid levels, respectively. WNT10B expression was not observed in normal and benign proliferations of human breast tissue but was found to be elevated in 3 of 50 primary breast carcinomas. Southern blot analysis of the carcinoma expressing the highest level of WNT10B showed no amplification or rearrangement of the gene.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.