Order Entry
United States
ContactUsLinkComponent
Human TNFSF13 ELISA Kit
Human TNFSF13 ELISA Kit
Catalog # MSPP-SEB750HU
CAS Number:  
undefined
Human TNFSF13 ELISA Kit
Catalog # MSPP-SEB750HU
Supplier Number:  SEB750HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    TNFSF13
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Tumor Necrosis Factor Ligand Superfamily, Member 13 (TNFSF13) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    46.88 - 3000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    17.74 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB750HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human TNFSF13 (Tumor Necrosis Factor Ligand Superfamily, Member 13). The assay range is from 46.88 to 3000 pg/ml (Sandwich kit) with a sensitivity of 17.74 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Tumor necrosis factor ligand superfamily member 13 is a member of the tumor necrosis factor (TNF) ligand family. Expressed at high levels in transformed cell lines, cancers of colon, thyroid, lymphoid tissues and specifically expressed in monocytes and macrophages. This protein is a ligand for TNFRSF17/BCMA, a member of the TNF receptor family. This protein and its receptor are both found to be important for B cell development. in vitro experiments suggested that this protein may be able to induce apoptosis through its interaction with other TNF receptor family proteins such as TNFRSF6/FAS and TNFRSF14/HVEM. Three alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.