Order Entry
United States
ContactUsLinkComponent
Human TGFb1 ELISA Kit
Human TGFb1 ELISA Kit
Catalog # MSPP-SEA124HU
CAS Number:  
undefined
Human TGFb1 ELISA Kit
Catalog # MSPP-SEA124HU
Supplier Number:  SEA124HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    TGFb1
  • Size:
    1 kit
  • Sample Type:
    Serum, platelet-poor plasma, tissue homogenates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Transforming Growth Factor Beta 1 (TGFb1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    15.6 - 1000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    5.7 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA124HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human TGFb1 (Transforming Growth Factor Beta 1). The assay range is from 15.6 to 1000 pg/ml (Sandwich kit) with a sensitivity of 5.7 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

TGF-β is capable of producing a variety of effects and virtually all cell types respond to this factor in some way. The inappropriate presence of active TGF-β1 has been implicated in a variety of pathological conditions Because of the necessity for regulating its activity tightly, TGF-β is secreted by cells in the form of an inactive complex. This complex consists of TGF-β1 associated non-covalently with a protein designated the latency associated peptide (LAP). TGF-β1 and LAP represent components of a pro-peptide that is cleaved in a post-golgi compartment prior to secretion. LAP and TGF-β1 each consist of a disulfide-linked homodimer and the association of these two components renders TGF-β1 inactive and inaccessible to anti-TGF-β antibodies.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.