- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:SMOC1
- Size:1 kit
- Sample Type:Serum, plasma, tissue homogenates and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between SPARC Related Modular Calcium Binding Protein 1 (SMOC1) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:0.156 - 10 ng/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:0.056 ng/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEN564HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human SMOC1 (SPARC Related Modular Calcium Binding Protein 1). The assay range is from 0.156 to 10 ng/ml (Sandwich kit) with a sensitivity of 0.056 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
The mature 408-amino acid protein contains a 26-residue signal peptide at the N terminus, followed by a follistatin-like domain, 2 thyroglobulin-like domains interrupted by a novel domain, and an extracellular calcium-binding domain. Recombinant expression in human cells showed that SMOC1 is a glycoprotein with a calcium-dependent conformation. Circular dichroism spectroscopy showed that SMOC1 has an approximately 40% beta-sheet structure. Northern blot analysis of adult mouse tissues revealed a 3.7-kb transcript with strong expression in ovary, and lower expression in brain, thymus, heart, skeletal muscle, liver, and lung. RT-PCR analysis detected a signal in testis also. Immunofluorescence studies showed that SMOC1 localized to basement membranes both in cultured cells and in mouse skin, kidney, and ovary.