Order Entry
United States
ContactUsLinkComponent
Human GR ELISA Kit
Human GR ELISA Kit
Catalog # MSPP-SEB314HU
CAS Number:  
undefined
Human GR ELISA Kit
Catalog # MSPP-SEB314HU
Supplier Number:  SEB314HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    GR
  • Size:
    1 kit
  • Sample Type:
    Tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Glutathione Reductase (GR) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    1.56 - 100 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.60 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEB314HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human GR (Glutathione Reductase). The assay range is from 1.56 to 100 ng/ml (Sandwich kit) with a sensitivity of 0.60 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Glutathione reductase, also known as GSR or GR, is an enzyme (EC 1.8.1.7) that reduces glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. For every mole of oxidized glutathione (GSSG), one mole of NADPH is required to reduce GSSG to GSH. The enzyme forms a FAD bound homodimer. The glutathione reductase is conserved between all kingdoms. In bacteria, yeasts, and animals, one glutathione reductase gene is found; however, in plant genomes, two GR genes are encoded. Drosophila and Trypanosomes do not have any GR at all. In these organisms, glutathione reduction is performed by either the thioredoxin or the trypanothione system, respectively.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.