Order Entry
United States
ContactUsLinkComponent
Human CTGF ELISA Kit
Human CTGF ELISA Kit
Catalog # MSPP-SEA010HU
CAS Number:  
undefined
Human CTGF ELISA Kit
Catalog # MSPP-SEA010HU
Supplier Number:  SEA010HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    CTGF
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Connective Tissue Growth Factor (CTGF) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.78 - 50 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.32 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA010HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human CTGF (Connective Tissue Growth Factor). The assay range is from 0.78 to 50 ng/ml (Sandwich kit) with a sensitivity of 0.32 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

CTGF (connective tissue growth factor) is a cysteine-rich, matrix-associated, heparin-binding protein. in vitro, CTGF mirrors some of the effects of TGF beta on skin fibroblasts, such as stimulation of extracellular matrix production, chemotaxis, proliferation and integrin expression. CTGF can promote endothelial cell growth, migration, adhesion and survival and is thus implicated in endothelial cell function and angiogenesis. CTGF binds to perlecan, a proteoglycan which has been localised in synovium, cartilage and numerous other tissues.CTGF has been implicated in extracellular matrix remodelling in wound healing, scleroderma and other fibrotic processes, as it is capable of upregulating both matrix metalloproteinases and their inhibitors.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.