- Assay duration:Multiple steps
- Assay Type:Sandwich
- Conjugate ELISA:Biotin
- Format:Pre-coated
- Host:Rabbit
- Primary antibody reactivity:Human
- Target protein:CD10
- Size:1 kit
- Sample Type:Serum, plasma, cerebrospinal fluid and other biological fluids
- Cross Reactivity:No significant cross-reactivity or interference between Neprilysin (CD10) and analogues was observed
- Detection Method:Colorimetric
- Time to Results:3 h
- Shelf Life:12 Months
- Detection Range:31.2 - 2000 pg/ml
- Storage Temperature:4 °C for one month (frequent use), −20 °C for one year
- Sample Volume:100 µl
- Sensitivity:12.5 pg/ml
- Regulatory Status:RUO
- Cat. No.:MSPP-SEB785HU
- No. of tests:96 wells
This assay has high sensitivity and excellent specificity for detecting Human CD10 (Neprilysin). The assay range is from 31.2 to 2000 pg/ml (Sandwich kit) with a sensitivity of 12.5 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.
- High sensitivity and specificity
- Perfect reproducibility and consistency across batches
- Quality control with three-level inspections
- Wide range of targets/species available
- Intra-assay: CV<10%; Inter-assay: CV<12%
Neprilysin is a zinc-dependent metalloprotease enzyme that degrades a number of small secreted peptides, most notably the amyloid beta peptide whose abnormal misfolding and aggregation in neural tissue has been implicated as a cause of Alzheimer's disease. Synthesized as a membrane-bound protein, the neprilysin ectodomain is released into the extracellular domain after it has been transported from the Golgi apparatus to the cell surface.
In neurons, neprilysin is regulated by the protein nicastrin, a component of the gamma secretase complex that performs a necessary step in processing amyloid precursor protein to amyloid beta.Neprilysin is also associated with other biochemical processes, and is particularly highly expressed in kidney and lung tissues.