Order Entry
United States
ContactUsLinkComponent
Human C1qA ELISA Kit
Human C1qA ELISA Kit
Catalog # MSPP-SED207HU
Supplier:  Cloud-Clone
CAS Number:  
undefined
Human C1qA ELISA Kit
Catalog # MSPP-SED207HU
Supplier:  Cloud-Clone
Supplier Number:  SED207HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    C1qA
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Complement Component 1, Q Subcomponent A (C1qA) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    78 - 5000 pg/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    35 pg/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SED207HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human C1qA (Complement Component 1, Q Subcomponent A). The assay range is from 78 to 5000 pg/ml (Sandwich kit) with a sensitivity of 35 pg/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Reid (1974) reported a partial amino acid sequence of 95 residues of the 191 residues in the oxidized A chain of human subcomponent C1q. This region of the A chain contains a repeating sequence of glycine-X-Y, where X is often proline and Y is often hydroxyproline, for 78 residues. The 5 hydroxylysine residues and 5 hydroxyproline residues in the oxidized A chain are all in these 78 residues and only in the Y position of the repeating sequence. Prolonged collagenase digestion of the oxidized A chain yielded a large, apparently C-terminal peptide containing most of the noncollagenous sequences present in the chain. Reid (1974) concluded that the A chain of C1q contains a collagen-like region that constitutes most of the N-terminal half of the chain.

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.