Order Entry
United States
ContactUsLinkComponent
Human ANGPT1 ELISA Kit
Human ANGPT1 ELISA Kit
Catalog # MSPP-SEA008HU
CAS Number:  
undefined
Human ANGPT1 ELISA Kit
Catalog # MSPP-SEA008HU
Supplier Number:  SEA008HU
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Host:
    Rabbit
  • Primary antibody reactivity:
    Human
  • Target protein:
    ANGPT1
  • Size:
    1 kit
  • Sample Type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross Reactivity:
    No significant cross-reactivity or interference between Angiopoietin 1 (ANGPT1) and analogues was observed
  • Detection Method:
    Colorimetric
  • Time to Results:
    3 h
  • Shelf Life:
    12 Months
  • Detection Range:
    0.24 - 15 ng/ml
  • Storage Temperature:
    4 °C for one month (frequent use), −20 °C for one year
  • Sample Volume:
    100 µl
  • Sensitivity:
    0.11 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    MSPP-SEA008HU
  • No. of tests:
    96 wells

Specifications

About this item

This assay has high sensitivity and excellent specificity for detecting Human ANGPT1 (Angiopoietin 1). The assay range is from 0.24 to 15 ng/ml (Sandwich kit) with a sensitivity of 0.11 ng/ml. There is no detectable cross-reactivity with other relevant proteins. Activity loss rate and accelerated stability test ect have been conducted to guarantee the best performance of the products after long storage and delivery.

  • High sensitivity and specificity
  • Perfect reproducibility and consistency across batches
  • Quality control with three-level inspections
  • Wide range of targets/species available
  • Intra-assay: CV<10%; Inter-assay: CV<12%

Angiopoietin 1 is a type of angiopoietin and is encoded by the gene ANGPT1. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme. The protein also contributes to blood vessel maturation and stability, and may be involved in early development of the heart. Angiopoietin 1 has been shown to interact with TEK tyrosine kinase. Angiopoietin1 (Ang1) is a novel angiogenic factor with important actions on endothelial cell (EC) differentiation and vascular maturation. Ang1 has been shown to prevent EC apoptosis through activation of PI3-kinase/Akt, a pathway that is also known to activate endothelium nitric oxide synthase (eNOS).

Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.