Extension Thermometer Clamp
Extention thermometer clamp
Catalog # 470357-310
Supplier: United Scientific Supplies
Supplier #: EXTHCL01
Jump to:
Product Details & Documents
Hold thermometers at odd angles and distances safety.
- All stainless steel construction
- Holds glass thermometers firmly but gently
- Easily adjust tension with the included wing nut
This extension clamp holds glass tubing or thermometers securely but gently. Adjustments are made easily with one hand using the wing nut. Clamp opening extends from 6 to 16 mm. Jaw width is 15.5 mm. Includes a stainless steel rod that is 15 cm long with a diameter of 10 mm. Overall length is 20 cm. Requires a right-angle clamp to connect to a support rod.
Specifications
- Description:Extention thermometer clamp