Order Entry
United States
ContactUsLinkComponent
Multiple Species Recombinant Cor (from Synthetic), BSA (Bovine Serum Albumin)
Multiple Species Recombinant Cor (from Synthetic), BSA (Bovine Serum Albumin)
Catalog # MSPP-CPA067GE1
Supplier:  Cloud-Clone
CAS Number:  
undefined
Multiple Species Recombinant Cor (from Synthetic), BSA (Bovine Serum Albumin)
Catalog # MSPP-CPA067GE1
Supplier:  Cloud-Clone
Supplier Number:  CPA067GE11
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    BSA (Bovine Serum Albumin)
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Synthetic
  • Species:
    Multiple
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Cortisone
  • Protein/Peptide Name:
    Cor
  • Purity:
    90 - 100%
  • Molecular Weight:
    360.4Da
  • Sequence:
    C21H28O5
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-CPA067GE1

Specifications

About this item

This is a Cor conjugated BSA, General species is sequencing from C21H28O5 with 90 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Cortisone is a steroid hormone. Chemically, it is a corticosteroid closely related to corticosterone. It is used to treat a variety of ailments and can be administered intravenously, orally, intraarticularly, or cutaneously. Cortisone suppresses the immune system, thus reducing inflammation and attendant pain and swelling at the site of the injury. However, its effects on the immune system have resulted in various side-effects, particularly in the long-term use of cortisone. 17-hydroxycorticosteroid (17-OHCS) is an inactive product formed when the liver and other body tissues break down cortisol, a steroid hormone.17-OHCS is a group of hydroxycorticosteroids bearing a hydroxy group at the 17-position. Urinary excretion of these compounds is used as an index of adrenal function. They are used systemically in the free alcohol form, but with esterification of the hydroxy groups, topical effectiveness is increased.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.