Order Entry
United States
ContactUsLinkComponent
Human TRAIL Active (Prokaryotic) (from E. coli)
Human TRAIL Active (Prokaryotic) (from E. coli)
Catalog # MSPP-APA139HU1
CAS Number:  
undefined
Human TRAIL Active (Prokaryotic) (from E. coli)
Catalog # MSPP-APA139HU1
Supplier Number:  APA139HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokines and Growth Factors
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    8743
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Tumor Necrosis Factor Related Apoptosis Inducing Ligand
  • UniProtKB:
    P50591
  • Protein/Peptide Name:
    TRAIL Active
  • Purity:
    95 - 100%
  • Molecular Weight:
    19 kDa
  • Sequence:
    Val114~Gly281
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Cell culture, Activity Assays.
  • Cat. No.:
    MSPP-APA139HU1

Specifications

About this item

This is a TRAIL active protein (prokaryotic), Human is sequencing from Val114~Gly281 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

TRAIL is a recently identified member of the TNF gene superfamily. Five different receptors have been identified for TRAIL. Two receptors, DR4 and DR5, are transmembrane proteins containing death domain similar to FAS and other TNF family receptors. Two other receptors, DcR1 and DcR2, act like decoy proteins for TRAIL binding because they lack the death domain. TRAIL can also bind, though weakly, to osteoprotegrin (OPG), a soluble receptor, which plays a role in osteoclastogenesis. TRAIL induces apoptosis in various tumor cell lines, whereas most primary cells seem to be resistant. TRAIL-mediated apoptosis occurs following its binding to DR4 or DR5 receptors. The mechanism of apoptosis involves activation of caspase-8 and subsequent activation of effector caspases.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.