Order Entry
United States
ContactUsLinkComponent
Human Recombinant ACE2 (Prokaryotic) (from E. coli)
Human Recombinant ACE2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB886HU1
CAS Number:  
undefined
Human Recombinant ACE2 (Prokaryotic) (from E. coli)
Catalog # MSPP-RPB886HU1
Supplier Number:  RPB886HU01
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    59272
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Angiotensin I Converting Enzyme 2
  • UniProtKB:
    Q9BYF1
  • Protein/Peptide Name:
    ACE2
  • Purity:
    95 - 100%
  • Molecular Weight:
    44 kDa
  • Sequence:
    Leu392~Ser740
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPB886HU1

Specifications

About this item

This is a ACE2 recombinant protein (prokaryotic), Human is sequencing from Leu392~Ser740 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

ACE2 is an exopeptidase that catalyses the conversion of angiotensin I to the nonapeptide angiotensin, or the conversion of angiotensin II to angiotensin. ACE 2 has direct effects on cardiac function, and is expressed predominantly in vascular endothelial cells of the heart and the kidneys.By EST database searching for sequences showing homology to the zinc metalloprotease angiotensin-I converting enzyme (ACE) and by screening a human lymphoma cDNA library, Tipnis et al. (2000) cloned a full-length ACE2 cDNA, which they called ACEH, encoding a deduced 805-amino acid protein that shares approximately 40% identity with the N- and C-terminal domains of ACE. ACE2 contains a potential 17-amino acid N-terminal signal peptide and a putative 22-amino acid C-terminal membrane anchor.

Certifications: ISO 9001 and ISO 13485 certified, three level QC system.
Caution: For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.