Order Entry
United States
ContactUsLinkComponent
 
Human Recombinant MUC5B (Prokaryotic) (from E. coli)
  MSPP-RPA684HU1
 :  
undefined
Human Recombinant MUC5B (Prokaryotic) (from E. coli)
  MSPP-RPA684HU1
 :  RPA684HU01
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    50 µg
  • Tag sequence:
    MGHHHHHHSGSEF
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    <1.0 EU per 1 μg (determined by the LAL method)
  • Gene ID:
    727897
  • Reconstitution Instructions:
    Reconstitute in 10 mM PBS (pH 7.4) to a concentration of 0.1 - 1.0 mg/ml. Do not vortex.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Mucin 5 Subtype B
  • UniProtKB:
    Q9HC84
  • Protein/Peptide Name:
    MUC5B
  • Purity:
    95 - 100%
  • Molecular Weight:
    17 kDa
  • Sequence:
    Lys5366~Val5444
  • Endotoxin Level:
    Low
  • Concentration:
    0.2 mg/ml
  • Formulation:
    Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose.
  • Nuclease-free:
    N
  • Shipping Temperature:
    4 °C
  • Tested Applications:
    Positive control, Immunogen, SDS-PAGE, Western blot.
  • Cat. No.:
    MSPP-RPA684HU1

 

 

This is a MUC5B recombinant protein (prokaryotic), Human is sequencing from Lys5366~Val5444 with 95 - 100% purity. Lyophilized from PBS, pH 7.4, containing 0.01% SKL, 5% Trehalose with 0.2 mg/ml.

  • High quality, purity, reproducibility and effectiveness
  • Offers customized buffers and tag options
  • 100% quality and service satisfaction guarantee

Mucins are high molecular mass, highly glycosylated macromolecules that are the major components of mucus secretions. MUC5B is a salivary mucin that is thought to contribute to the lubricating and viscoelastic properties of whole saliva. It is composed of 14.9% protein, 78.1% carbohydrate, and 7% sulfate.Desseyn et al. isolated MUC5B genomic sequence. They found that the central exon encodes a predicted 3,570-amino acid peptide containing 19 subdomains that can be grouped into 4 larger composite units referred to as 'superrepeats'. Since MUC5B and the related proteins MUC2 and MUC5AC are clustered within chromosome 11p15.5, suggesting that they are members of a multigene family.Desseyn et al. reported that the region 3-prime of the large central exon of MUC5B is composed of 18 exons and spans about 10.7 kb.

 : ISO 9001 and ISO 13485 certified, three level QC system.
 : For research use only. Not for use in clinical diagnostic procedures. Please proper stored each component based on the instruction.